Merck
Caffeine intake antagonizes salt sensitive hypertension through improvement of renal sodium handling.
Scientific reports (2016-05-14)
Hao Yu, Tao Yang, Peng Gao, Xing Wei, Hexuan Zhang, Shiqiang Xiong, Zongshi Lu, Li Li, Xiao Wei, Jing Chen, Yu Zhao, William J Arendshorst, Qianhui Shang, Daoyan Liu, Zhiming Zhu
ABSTRAKT
High salt intake is a major risk factor for hypertension. Although acute caffeine intake produces moderate diuresis and natriuresis, caffeine increases the blood pressure (BP) through activating sympathetic activity. However, the long-term effects of caffeine on urinary sodium excretion and blood pressure are rarely investigated. Here, we investigated whether chronic caffeine administration antagonizes salt sensitive hypertension by promoting urinary sodium excretion. Dahl salt-sensitive (Dahl-S) rats were fed with high salt diet with or without 0.1% caffeine in drinking water for 15 days. The BP, heart rate and locomotor activity of rats was analyzed and urinary sodium excretion was determined. The renal epithelial Na(+) channel (ENaC) expression and function were measured by in vivo and in vitro experiments. Chronic consumption of caffeine attenuates hypertension induced by high salt without affecting sympathetic nerve activity in Dahl-S rats. The renal α-ENaC expression and ENaC activity of rats decreased after chronic caffeine administration. Caffeine increased phosphorylation of AMPK and decrease α-ENaC expression in cortical collecting duct cells. Inhibiting AMPK abolished the effect of caffeine on α-ENaC. Chronic caffeine intake prevented the development of salt-sensitive hypertension through promoting urinary sodium excretion, which was associated with activation of renal AMPK and inhibition of renal tubular ENaC.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Inhibitor AMPK, związek C, AMPK Inhibitor, Compound C, CAS 866405-64-3, is a cell-permeable compound that inhibits KDR/VEGFR2, ALK2/BMPR-I, and AMPK kinase activities (IC50 = 25.1, 148, and 234.6 nM, respectively).
Sigma-Aldrich
Anti-Thiazide-Sensitive NaCl Cotransporter Antibody, Chemicon®, from rabbit