Merck
Self-associating cellulose-graft-poly(ε-caprolactone) to design nanoparticles for drug release.
Materials science & engineering. C, Materials for biological applications (2020-01-12)
Simona Zuppolini, Iriczalli Cruz Maya, Laura Diodato, Vincenzo Guarino, Anna Borriello, Luigi Ambrosio
ABSTRAKT
The growing interest in the use of polysaccharides nanoparticles for biomedical applications is related to the recent progresses on the synthesis of cellulose-based polymers with the specific functionalities. In particular, cellulose graft copolymers are emerging as amphiphilic materials suitable to fabricate smart nanoparticles for drug delivery applications. In this work, a cellulose-graft-poly(ε-caprolactone) (cell-g-PCL) was synthetized and characterized by FTIR, TGA and DSC in order to validate the synthesis process. We demonstrated that fast evaporation processes promoted cell-g-PCL self-assembly to form nanomicellar structures with hydrodynamic radius ranged from 30 to 60 nm as confirmed by TEM analysis. Moreover, the application of controlled electrostatic forces on solvent evaporation - namely electrospraying - allowed generating round-like nanoscaled particles, as confirmed by SEM supported via image analysis. We demonstrated also that sodium diclofenac (DS) drastically influenced the mechanism of particle formation, favoring the deposition of erythrocyte-like particles with highly concave surfaces, not penalizing the encapsulation efficiency of nanoparticles (>80%). The release profile showed a fast delivery of DS - about 60% during the first 24 h - followed by a sustained release - about 20% during the next 6 days - strictly related to the peculiar weak interactions among amphiphilic polymer segments and water molecules, thus suggesting a successful use of electrosprayed cell-g-PCL nanoparticles for therapeutic treatments in nanomedicine.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,1,1,3,3,3-Hexafluoro-2-propanol, ≥99%