Merck
Effect of enzymatically hydrolyzed yeast on health and performance of transition dairy cattle.
Journal of dairy science (2019-12-23)
H Stefenoni, J H Harrison, A Adams-Progar, E Block
ABSTRAKT
The objective of this study was to evaluate the effect of supplementing enzymatically hydrolyzed yeast (EHY; Celmanax Arm and Hammer Animal Nutrition, Princeton, NJ) on transition dairy cattle. Forty multiparous Holstein cows were blocked by predicted transmitting ability and randomly assigned to 1 of 2 treatments (EHY, n = 20; or control, CON, n = 20) from 21 d before expected calving to 60 d postpartum. The EHY cows received 56 and 28 g/d in close-up and lactating diets, respectively. Dry matter intake, health events, milk production parameters, feed efficiency, colostrum quality, reproductive parameters, body weight, and body condition score were monitored. Fecal samples collected on -21, -14, -7, 0, 1, 3, 5, 7, and 14 d relative to calving were analyzed for total coliforms, Clostridium perfringens, Salmonella, and Escherichia coli O157:H7. Blood samples were collected at 7, 14, and 21 d postpartum for analysis of β-hydroxybutyrate. Sterile quarter milk samples collected at dry-off, calving, and wk 1, 2, and 3 of lactation were analyzed for milk pathogens and somatic cell count. Pre- or postpartum dry matter intake, body weight, body condition score, milk yield, and milk protein and fat yields did not differ among treatments. Milk fat and protein concentrations were greater in EHY cows than CON cows. β-Hydroxybutyrate and health events were not different among treatments. The presence of fecal C. perfringens did not differ prepartum, but was lower in EHY cows postpartum. Milk pathogens and total intramammary infections did not differ between treatments at dry-off, calving, wk 1, or wk 2, but more EHY cows were infected with Staphylococcus sp. during wk 3 than CON cows. The EHY cows showed heat earlier than CON cows, but no other reproductive parameters were affected. The EHY supplementation during the transition period did not affect dry matter intake, milk yield, health events, or reproductive parameters but did increase milk protein and fat concentrations.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, 98%