Merck
METTL3 regulates inflammatory pain by modulating m6A-dependent pri-miR-365-3p processing.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-01-10)
Chenjing Zhang, Yin Wang, Yunan Peng, Hongjiao Xu, Xuelong Zhou
ABSTRAKT
N6-methyladenosine (m6A) modification in RNA has been implicated in diverse biological processes. However, very little is currently known about its role in nociceptive modulation. Here, we found that the level of spinal m6A modification was significantly increased in a mouse model of Complete Freund's Adjuvant (CFA)-induced chronic inflammatory pain, which was accompanied with the augmentation of methyltransferase-like 3 (METTL3) expression in the spinal cord. Knockdown of spinal METTL3 prevented and reversed CFA-induced pain behaviors and spinal neuronal sensitization. In contrast, overexpression of spinal METTL3 produced pain behaviors and neuronal sensitization in naive mice. Moreover, we found that METTL3 positively modulated the pri-miR-65-3p processing in a microprocessor protein DiGeorge critical region 8-dependent manner. Collectively, our findings reveal an important role of METTL3-mediated m6A modification in nociceptive sensitization and provide a novel perspective on m6A modification in the development of pathological pain.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Formalin solution, neutral buffered, 10%, case of 48 × 15 mL, histological tissue fixative
Sigma-Aldrich
GenElute mRNA Miniprep Kit, sufficient for 70 purifications
Sigma-Aldrich
Freund′s Adjuvant, Complete, cell suspension
Sigma-Aldrich
Zestaw do immunoprecypitacji białek wiążących RNA Magna RIP®, RNA Immunoprecipitation (RIP) Kit containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions using protein A/G magnetic beads.