Merck
Beclin 1 Promotes Endosome Recruitment of Hepatocyte Growth Factor Tyrosine Kinase Substrate to Suppress Tumor Proliferation.
Cancer research (2019-11-21)
Asia N Matthew-Onabanjo, Jenny Janusis, Jose Mercado-Matos, Anne E Carlisle, Dohoon Kim, Fayola Levine, Peter Cruz-Gordillo, Ryan Richards, Michael J Lee, Leslie M Shaw
ABSTRAKT
Beclin 1 has nonautophagic functions that include its ability to regulate endocytic receptor trafficking. However, the contribution of this function to tumor suppression is poorly understood. Here, we provide in vivo evidence that Beclin 1 suppresses tumor proliferation by regulating the endocytic trafficking and degradation of the EGFR and transferrin (TFR1) receptors. Beclin 1 promoted endosomal recruitment of hepatocyte growth factor tyrosine kinase substrate (HRS), which was necessary for sorting surface receptors to intraluminal vesicles for signal silencing and lysosomal degradation. In tumors with low Beclin 1 expression, endosomal HRS recruitment was diminished and receptor function was sustained. Collectively, our results demonstrate a novel role for Beclin 1 in impeding tumor growth by coordinating the regulation of key growth factor and nutrient receptors. These data provide an explanation for how low levels of Beclin 1 facilitate tumor proliferation and contribute to poor cancer outcomes. SIGNIFICANCE: Beclin 1 controls the trafficking fate of growth regulatory receptors to suppress tumor proliferation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-phospho-HRS (pTyr334) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Lapatinib
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder