Merck
Light alters the responses of two marine diatoms to increased warming.
Marine environmental research (2020-01-14)
Xiaopeng Zeng, Peng Jin, Yingying Jiang, Haimei Yang, Jiahui Zhong, Zhe Liang, Yingyan Guo, Peiyuan Li, Quanting Huang, Jinmei Pan, Hua Lu, Yanyun Wei, Dinghui Zou, Jianrong Xia
ABSTRAKT
In this study, we examined the effects of increased temperature (15, 20 and 25 °C) and different light levels (50, 200 μmol photons m-2 s-1) on two widely distributed diatoms, namely Phaeodactylum tricornutum and Thalassiosira weissflogii. Results showed that increasing light level counteracted the negative effects of high temperature on photosynthesis in both species, suggesting an antagonistic interaction between light and temperature. Contrary to the above results, light limitation diminished the temperature-sensitivity of carbonic anhydrase activity in two diatoms. We also observed species-specific responses of biomass, where increased temperature significantly decreased the biomass of P. tricornutum at both low and high light levels but showed no effects on T. weissflogii. Our study demonstrated that light can alter the physiological responses of diatoms to temperature but also revealed interspecific variations. We predict that in the future ocean with shallower upper mixed layer, T. weissflogii may be more competitive than P. tricornutum.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O