Merck
Bcl2l10 mediates the proliferation, invasion and migration of ovarian cancer cells.
International journal of oncology (2020-01-03)
Su-Yeon Lee, Jinie Kwon, Ji Hye Woo, Kyeoung-Hwa Kim, Kyung-Ah Lee
ABSTRAKT
Bcl2l10, also known as Diva, Bcl‑b and Boo, is a member of the Bcl2 family of proteins, which are involved in signaling pathways that regulate cell apoptosis and autophagy. Previously, it was demonstrated that Bcl2l10 plays a crucial role in the completion of oocyte meiosis and is a key regulator of Aurora kinase A (Aurka) expression and activity in oocytes. Aurka is overexpressed in several types of solid tumors and has been considered a target of cancer therapy. Based on these previous results, in the present study, the authors aimed to investigate the regulatory role of Bcl2l10 in A2780 and SKOV3 human ovarian cancer cells. The protein expression of Bcl2l10 was examined in human cancer tissues and cell lines, including the ovaries, using a tissue microarray and various human ovarian cancer cell lines. It was found that Bcl2l10 regulated the protein stability and activities of Aurka in ovarian cancer cells. Although apoptosis was not affected, the cell cycle was arrested at the G0/G1 phase by Bcl2l10 knockdown. Of note, cell viability and motility were markedly increased by Bcl2l10 knockdown. On the whole, the findings of this study suggest that Bcl2l10 functions as tumor suppressor gene in ovarian cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Bcl2l10