Merck
miR-324-5p upregulation potentiates resistance to cisplatin by targeting FBXO11 signalling in non-small cell lung cancer cells.
Journal of biochemistry (2019-11-30)
Zhichang Ba, Yufei Zhou, Zhaoyang Yang, Jianyu Xu, Xiushi Zhang
ABSTRAKT
Dysregulation of microRNAs (miRNAs) plays a key role during the pathogenesis of chemoresistance in lung cancer (LCa). Previous study suggests that miR-324-5p may serve as a unique miRNA signature for LCa, but its role and the corresponding molecular basis remain largely explored. Herein, we report that miR-324-5p expression was significantly increased in cisplatin (CDDP)-resistant LCa tissues and cells, and this upregulation predicted a poor post-chemotherapy prognosis in LCa patients. miR-324-5p was further shown to impact CDDP response: Ectopic miR-324-5p expression in drug-naïve LCa cells was sufficient to attenuate sensitivity to CDDP and to confer more robust tumour growth in CDDP-challenged nude mice. Conversely, ablation of miR-324-5p expression in resistant cells effectively potentiated CDDP-suppressed cell growth in vitro and in vivo. Using multiple approaches, we further identified the tumour suppressor FBXO11 as the direct down-stream target of miR-324-5p. Stable expression of FBXO11 could abrogate the pro-survival effects of miR-324-5p in CDDP-challenged LCa cells. Together, these findings suggest that miR-324-5p upregulation mediates, at least partially, the CDDP resistance by directly targeting FBXO11 signalling in LCa cells. In-depth elucidation of the molecular basis underpinning miR-324-5p action bears potential implications for mechanism-based strategies to improve CDDP responses in LCa.