Merck
Pro-survival and anti-inflammatory roles of NF-κB c-Rel in the Parkinson's disease models.
Redox biology (2020-01-28)
Zishan Wang, Hongtian Dong, Jinghui Wang, Yulu Huang, Xiaoshuang Zhang, Yilin Tang, Qing Li, Zhaolin Liu, Yuanyuan Ma, Jiabin Tong, Li Huang, Jian Fei, Mei Yu, Jian Wang, Fang Huang
ABSTRAKT
The pathological hallmarks of Parkinson's disease (PD) are the progressive loss of dopaminergic (DA) neurons in the substantia nigra pars compacta (SNpc) and the presence of overactivated glial cells and neuroinflammation. Nuclear factor kappa-light-chain-enhancer of activated B cells (NF-κB) c-Rel subunit is closely related in the pathological progress of PD, however the roles and mechanisms of c-Rel in PD development remain unclear. Here, in neurotoxins-induced PD models, the dynamic changes of NF-κB c-Rel and its functions were evaluated. We found that c-Rel was rapidly activated in the nigrostriatal pathway, which mainly occurred in dopaminergic neurons and microglia. c-Rel could maintain neuronal survival by initiating the anti-apoptotic gene expression in MPP+-treated SH-SY5Y cells and it could inhibit microglial overactivation by suppressing the inflammatory gene expression in LPS-challenged BV2 cells. c-Rel inhibitor IT901 aggravated the damage of MPTP on dopaminergic neurons and promoted the activation of microglia in the nigrostriatal pathway of mice. Moreover, the expression of c-Rel in blood samples of PD patients decreased dramatically. Our results indicate that the NF-κB/c-Rel subunit plays an important role in neuroprotection and neuroinflammation inhibition during PD progression.