Merck
  • Chemical and physical reinforcement behavior of dialdehyde nanocellulose in PVA composite film: A comparison of nanofiber and nanocrystal.
Chemical and physical reinforcement behavior of dialdehyde nanocellulose in PVA composite film: A comparison of nanofiber and nanocrystal.
Carbohydrate polymers (2020-01-19)
Hohyun Lee, Jinhwa You, Hyoung-Joon Jin, Hyo Won Kwak
ABSTRAKT
Nanocellulose can be categorized into cellulose nanofibers (CNFs) and cellulose nanocrystals (CNCs). CNFs and CNCs are oxidized by periodate oxidation to introduce dialdehyde groups, which can act as a crosslinking agent. Polyvinyl alcohol (PVA) is widely used in food packaging, however, it is vulnerable to aqueous environments. In this study, dialdehyde nanocellulose-reinforced PVA nanocomposite films were fabricated to enhance the water resistance. CNF and CNC were oxidized using varying amounts of sodium periodate and the selected fillers were designated as DCNF and DCNC, respectively. In the case of DCNF, physicochemical networking effect was induced to PVA, while DCNC showed only chemical network reinforcement effect. However, both PVA/DCNF and PVA/DCNC composite films have significantly improved vulnerabilities in moisture environment compared to PVA films. These results indicate that while physical network structures are important, the chemically formed network structures can play a crucial role in enhancing the water resistance of PVA films.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methyl Orange, ACS reagent, Dye content 85 %