Merck
Beta-1,3-glucan: a source of carbon and energy for chloroplast development in Euglena gracilis.
Australian journal of biological sciences (1971-02-01)
M R Dwyer, R M Smillie
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
β-1,3-Glucan from Euglena gracilis