Merck
  • The pro-inflammatory marker soluble suppression of tumorigenicity-2 (ST2) is reduced especially in diabetic morbidly obese patients undergoing bariatric surgery.
The pro-inflammatory marker soluble suppression of tumorigenicity-2 (ST2) is reduced especially in diabetic morbidly obese patients undergoing bariatric surgery.
Cardiovascular diabetology (2020-02-27)
Svitlana Demyanets, Christoph Kaun, Alexandra Kaider, Walter Speidl, Manfred Prager, Stanislav Oravec, Philipp Hohensinner, Johann Wojta, Gersina Rega-Kaun
ABSTRAKT
High soluble suppression of tumorigenicity-2 (sST2) is a marker of poor prognosis in chronic inflammatory conditions. ST2 and its ligand interleukin (IL)-33 are elevated in adipose tissue of obese individuals. We aimed to evaluate circulating sST2 and IL-33 as possible markers of metabolic benefit in morbidly overweight patients after Roux-en-Y gastric bypass (RYGB) bariatric surgery. sST2, IL-33, high sensitive IL-6, high sensitive C-reactive protein (hsCRP), leptin, cholesterol metabolism and liver parameters were measured in 80 morbidly obese individuals before and 1 year after bariatric surgery. sST2 was higher (P = 0.03) in diabetics as compared to individuals without diabetes. Baseline sST2 was also higher in males than in females (P= 0.0002). One year after bariatric surgery, sST2 levels were decreased (median 120, IQR 59-176 pg/mL) as compared to sST2 before surgery (median 141, IQR 111-181, P = 0.0024), and the diabetic group showed most pronounced reduction in sST2 (P = 0.0016). An association was found between sST2 and liver function parameters before and after bariatric surgery, and between baseline sST2 and total cholesterol, triglyceride, total low density lipoprotein (LDL), small dense LDL, Apolipoprotein B as well as with small dense high density lipoproteins (HDL). In the subgroup of diabetic patients positive correlation between IL-33 and sST2 (r = 0.44, P = 0.05) was noticed. Circulating sST2 is associated with markers of liver functions and lipid metabolism in severely obese patients and a reduction of sST2 was shown after successful bariatric surgery, most prominently in diabetic patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Human IL-33 ELISA Kit, for serum, plasma, cell culture supernatant and urine