Merck
  • Chaiqin chengqi decoction alleviates severe acute pancreatitis associated acute kidney injury by inhibiting endoplasmic reticulum stress and subsequent apoptosis.
Chaiqin chengqi decoction alleviates severe acute pancreatitis associated acute kidney injury by inhibiting endoplasmic reticulum stress and subsequent apoptosis.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2020-03-20)
Xuefei Yang, Xiaoxin Zhang, Ziqi Lin, Jia Guo, Xinmin Yang, Linbo Yao, Haoyang Wang, Ping Xue, Qing Xia
ABSTRAKT
Acute kidney injury (AKI), characterized by an increase of serum creatinine and urea, is a severe complication of severe acute pancreatitis (SAP) with high mortality. Endoplasmic reticulum (ER) stress has been considered as a key pathologic process in AKI. Chaiqin chengqi decoction (CQCQD) is an effective Chinese medicine formula for SAP treatment in China and has been used for many years. Our goal is to explore the role of CQCQD on ER stress of AKI in experimental SAP. SAP was induced in rats by retrograde duct injection of 5% sodium taurocholate (NaTC, 1 ml/kg), sham operation (SO) rats simultaneously received saline infusion. Intraperitoneal injection of 4-PBA (50 mg/kg, once a day for three days before the surgery) or intragastric gavage of CQCQD (1 g/kg, 2 hourly × 3 after disease induction) was used to treat SAP rats. All animals were humanely sacrificed 12 h after disease induction. Histopathology scores of kidney and pancreas; serum biochemical indices and kidney protein levels of ER stress and apoptosis markers were assessed. Tubular epithelial cell line (HK-2) was treated either with TNF-α (10 ng/ml) or IL-6 (10 ng/ml) for 12 h plus either 4-PBA (0.1 M) or CQCQD (1 mg/ml) for in vitro study. Cell viability and markers of ER stress and apoptosis were measured. Ductal perfusion of NaTC caused significant increases in serum lipase, amylase and pancreatic histopathology (inflammatory cell infiltration, interstitial edema, and acinar cell necrosis). Kidney histopathology (tubular dilation, brush border loss, little tubular necrosis, and cast formation), serum creatine and urea levels were raised when compared with the SO group. Moreover, apoptotic cell death markers (caspase-9, cleaved-caspase-3, and TUNEL) and kidney ER stress proteins (BIP, IRE1-α, XBP1s, and CHOP) were elevated after NaTC administration. 4-PBA and CQCQD significantly alleviated histopathological changes of kidney and pancreas, inflammatory cytokines, biochemical markers of AKI, ER stress proteins and apoptotic cell death markers. They also protected HK-2 cells from injury of TNF-α and IL-6, and alleviated both ER stress and apoptosis proteins in vitro. CQCQD may alleviate SAP-related AKI by inhibiting ER stress-related apoptosis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sodium taurocholate, BRP, European Pharmacopoeia (EP) Reference Standard