Merck
  • Patient and lesion characteristics in late/very late stent thrombosis with everolimus-eluting stents from real-world adverse event reporting.
Patient and lesion characteristics in late/very late stent thrombosis with everolimus-eluting stents from real-world adverse event reporting.
Journal of cardiology (2019-09-21)
Akihide Konishi, Yoshiaki Mitsutake, Mami Ho, Nobuhiro Handa, Kazuhisa Koike, Shuichi Mochizuki, Kensuke Ishii
ABSTRAKT
Stent thrombosis (ST) is a rare but serious complication after deployment of a drug-eluting stent. The features of ST after implantation of an everolimus-eluting stent (EES) have not been fully elucidated. A comprehensive survey of real-world adverse event reporting with conditions for approval under the Pharmaceuticals and Medical Devices Act identified 490 cases of ST with EES. A total of 370 patients with definite ST after EES implantation [287 with early (E)ST (within 30 days), 54 with late (L)ST (31-365 days), and 29 with very late (VL)ST (over 1 year)] were divided into an EST group and an LST/VLST group to compare the patients and lesions characteristics. The frequency of patients with hemodialysis and in-stent restenosis (ISR) lesions were significantly higher in the LST/VLST group than in the EST group (hemodialysis, 22.9% vs 2.7%, p = 0.0001; ISR lesions, 25.9% vs 9.4%, p = 0.0001). Characteristic demographic factors for LST/VLST versus EST identified by multivariable model were hemodialysis and ISR lesions (hemodialysis: odds ratio 7.348, 95% confidence interval 2.458-21.968, p = 0.0001; ISR lesions: odds ratio, 2.490, 95% confidence interval 1.100-5.638, p = 0.027). The in-hospital death rates from ST were not significantly different between the EST group and the LST/VLST group (EST, 15% vs LST/VLST, 21.7%, p = 0.147). Patient-related and lesion-related characteristics were significantly different between EST and LST/VLST. Data collection from adverse event reporting could be a helpful strategy for evaluation of this serious but rare complication.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)