- Daturmetesides A-E, five new ergostane-type C28 sterols from the leaves of Datura metel L.
Daturmetesides A-E, five new ergostane-type C28 sterols from the leaves of Datura metel L.
Steroids (2020-01-27)
Jin-Yan Tan, Yan Liu, Yan-Gang Cheng, Yan-Ping Sun, Yuan Liu, Jin Huang, Shuang Guo, Guo-Zhen Liu, Hai-Xue Kuang, Bing-You Yang
PMID31982422
ABSTRAKT
Five undescribed ergostane-type C28 sterols, daturmetesides A-E (1-5), were isolated from the leaves of Datura metel L. The chemical structures of these new compounds were characterized through extensive spectroscopic analysis and comparison with literatures. Among them, the absolute structures of daturmetesides A and C were unambiguously determined by X-ray crystallography. The anti-inflammatory effect of daturmetesides A-E was all tested by measuring nitric oxide production in lipopolysaccharide-activated RAW264.7 cells. Daturmetesides A, C and D moderatelylowered the NO production with IC50 values ranging from 17.05 ± 0.35 to 24.88 ± 0.93 μM.