- Stereoselective synthesis of L-guluronic acid alginates.
Stereoselective synthesis of L-guluronic acid alginates.
Chemistry (Weinheim an der Bergstrasse, Germany) (2008-09-05)
Jasper Dinkelaar, Leendert J van den Bos, Wouter F J Hogendorf, Gerrit Lodder, Herman S Overkleeft, Jeroen D C Codée, Gijsbert A van der Marel
PMID18770512
ABSTRAKT
The glycosylation properties of gulopyranosides have been mapped out, and it is shown that gulose has an intrinsic preference for the formation of 1,2-cis-glycosidic bonds. It is postulated that this glycosylation behaviour originates from nucleophilic attack at the oxacarbenium ion, which adopts the most favourable 3H4 conformation. Building on the stereoselectivity of gulose, a guluronic acid alginate trisaccharide was assembled for the first time by using gulopyranosyl building blocks.