Merck
A recurrent ABCC2 p.G693R mutation resulting in loss of function of MRP2 and hyperbilirubinemia in Dubin-Johnson syndrome in China.
Orphanet journal of rare diseases (2020-03-19)
Lina Wu, Yanmeng Li, Yi Song, Donghu Zhou, Siyu Jia, Anjian Xu, Wei Zhang, Hong You, Jidong Jia, Jian Huang, Xiaojuan Ou
ABSTRAKT
Dubin-Johnson syndrome (DJS) is a rare autosomal recessive disorder characterized by predominantly conjugated hyperbilirubinemia that is caused by pathogenic mutations in the adenosine triphosphate-binding cassette subfamily C member 2 (ABCC2) gene, which encodes multidrug resistance-associated protein 2 (MRP2). However, little is known about the causative mutation of DJS in China. Recently, we have reported ABCC2 p.G693R mutation in two unrelated cases. In the present study, we investigated the pathogenicity of the ABCC2 p.G693R mutation in DJS in China. Clinical and genetic analysis was conducted for the two patients with the ABCC2 p.G693R mutation. Whole exome sequencing for mutations in other known hyperbilirubinemia-related genes was conducted for the cases with ABCC2 p.G693R. Expression and cellular localization of the mutant MRP2 p.G693R were analyzed by Western blotting and immunofluorescence assay, respectively. Organic anion transport activity was evaluated by the analysis of glutathione-conjugated-monochlorobimane. The two DJS patients with ABCC2 p.G693R mutation, which was conserved among different species, showed typical hyperbilirubinemia phenotype. No pathogenic mutation was identified in the other known hyperbilirubinemia related genes. Functional studies in three cell lines showed that the expression, localization and the organic anion transport activity were significantly compromised by MRP2 p.G693R mutation compared with wild-type MRP2. The recurrent ABCC2 p.G693R mutation is associated with loss of function of the MRP2 protein and may result in hyperbilirubinemia in DJS in China.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phalloidin, Fluorescein Isothiocyanate Labeled, sequence Amanita phalloides(synthetic: peptide sequence)