Merck
  • Tungsten Nitride, Well-Dispersed on Porous Carbon: Remarkable Catalyst, Produced without Addition of Ammonia, for the Oxidative Desulfurization of Liquid Fuel.
Tungsten Nitride, Well-Dispersed on Porous Carbon: Remarkable Catalyst, Produced without Addition of Ammonia, for the Oxidative Desulfurization of Liquid Fuel.
Small (Weinheim an der Bergstrasse, Germany) (2019-06-06)
Nazmul Abedin Khan, Biswa Nath Bhadra, Suk Woo Park, Yong-Su Han, Sung Hwa Jhung
ABSTRAKT
Polyanilines (pANIs), loaded with phosphotungstic acid (PTA), are pyrolyzed to get WO3 or W2 N (≈6 and ≈7 nm, respectively), which is well-dispersed on pANI-derived porous carbons (pDCs). Depending on the pyrolysis temperature, WO3 /pDC, W2 N/pDC, or W2 N-W/pDCs could be obtained selectively. pANI acts as both the precursor of pDC and the nitrogen source for the nitridation of WO3 into W2 N during the pyrolysis. Importantly, W2 N could be obtained from the pyrolysis without ammonia feeding. The obtained W2 N/pDC is applied as a heterogeneous catalyst for the oxidative desulfurization (ODS) of liquid fuel for the first time, and the results are compared with WO3 /pDC and WO3 /ZrO2 . The W2 N/pDC is very efficient in ODS with remarkable performance compared with WO3 /pDC or WO3 /ZrO2 , which is applied as a representative ODS catalyst. For example, W2 N/pDC shows around 3.4 and 2.7 times of kinetic constant and turnover frequency (based on 5 min of reaction), respectively, compared to that of WO3 /ZrO2 . Moreover, the catalysts could be regenerated in a facile way. Therefore, W2 N/pDC could be produced facilely from pyrolysis (without ammonia feeding) of PTA/pANI, and W2 N, well-dispersed on pDC, can be suggested as a very efficient oxidation catalyst for the desulfurization of liquid fuel.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, 98%