Merck
  • A conductive cell-delivery construct as a bioengineered patch that can improve electrical propagation and synchronize cardiomyocyte contraction for heart repair.
A conductive cell-delivery construct as a bioengineered patch that can improve electrical propagation and synchronize cardiomyocyte contraction for heart repair.
Journal of controlled release : official journal of the Controlled Release Society (2020-01-21)
Shanglin Chen, Meng-Hsuan Hsieh, Shu-Hong Li, Jun Wu, Richard D Weisel, Yen Chang, Hsing-Wen Sung, Ren-Ke Li
ABSTRAKT
Cardiac tissue engineering is of particular importance in the combination of contracting cells with a biomaterial scaffold, which serves as a cell-delivery construct, to replace cardiomyocytes (CMs) that are lost as a result of an infarction, to restore heart function. However, most biomaterial scaffolds are nonconductive and may delay regional conduction, potentially causing arrhythmias. In this study, a conductive CM-delivery construct that consists of a gelatin-based gelfoam that is conjugated with a self-doped conductive polymer (poly-3-amino-4-methoxybenzoic acid, PAMB) is proposed as a cardiac patch (PAMB-Gel patch) to repair an infarcted heart. A nonconductive plain gelfoam (Gel patch) is used as a control. The electrical conductivity of the PAMB-Gel patch is approximately 30 times higher than that of the Gel patch; as a result, the conductive PAMB-Gel patch can substantially increase electrical conduction between distinct clusters of beating CMs, facilitating their synchronous contraction. In vivo epicardial implantation of the PAMB-Gel patch that is seeded with CMs (the bioengineered patch) in infarcted rat hearts can significantly enhance electrical activity in the fibrotic tissue, improving electrical impulse propagation and synchronizing CM contraction across the scar region, markedly reducing its susceptibility to cardiac arrhythmias. Echocardiography shows that the bioengineered conductive patch has an important role in the restoration of cardiac function, perhaps owing to the synergistic effects of its conductive construct and the synchronously beating CMs. These experimental results reveal that the as-proposed bioengineered conductive patch has great potential for repairing injured cardiac tissues.