Merck
Kirigami-Based Light-Induced Shape-Morphing and Locomotion.
Advanced materials (Deerfield Beach, Fla.) (2019-12-14)
Yu-Chieh Cheng, Hao-Chuan Lu, Xuan Lee, Hao Zeng, Arri Priimagi
ABSTRAKT
The development of stimuli-responsive soft actuators, a task largely undertaken by material scientists, has become a major driving force in pushing the frontiers of microrobotics. Devices made of soft active materials are oftentimes small in size, remotely and wirelessly powered/controlled, and capable of adapting themselves to unexpected hurdles. However, nowadays most soft microscale robots are rather simple in terms of design and architecture, and it remains a challenge to create complex 3D soft robots with stimuli-responsive properties. Here, it is suggested that kirigami-based techniques can be useful for fabricating complex 3D robotic structures that can be activated with light. External stress fields introduce out-of-plane deformation of kirigami film actuators made of liquid crystal networks. Such 2D-to-3D structural transformations can give rise to mechanical actuation upon light illumination, thus allowing the realization of kirigami-based light-fuelled robotics. A kirigami rolling robot is demonstrated, where a light beam controls the multigait motion and steers the moving direction in 2D. The device is able to navigate along different routes and moves up a ramp with a slope of 6°. The results demonstrate a facile technique to realize complex and flexible 3D structures with light-activated robotic functions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2,2-Dimethoxy-2-phenylacetophenone, 99%