Merck
An altered HLA-A0201-restricted MUC1 epitope that could induce more efficient anti-tumor effects against gastric cancer.
Experimental cell research (2020-03-12)
Huahui Yu, Chunmei Ye, Jieyu Li, Chunli Pan, Wansong Lin, Huijing Chen, Zhifeng Zhou, Yunbin Ye
ABSTRAKT
MUC1 is a tumor-associated antigen (TAA) overexpressed in many tumor types, which makes it an attractive target for cancer immunotherapy. However, this marker is a non-mutated antigen without high immunogenicity. In this study, we designed several new altered peptides by replacing amino acids in their sequences, which were derived from a low-affinity MUC1 peptide, thus bypassing immune tolerance. Compared to the wild-type (WT) peptide, the altered MUC1 peptides (MUC11081-1089L2, MUC11156-1164L2, MUC11068-1076Y1) showed higher affinity to the HLA-A0201 molecule and stronger immunogenicity. Furthermore, these altered peptides resulted in the generation of more cytotoxic T lymphocytes (CTLs) that could cross-recognize gastric cancer cells expressing WT MUC1 peptides, in an HLA-A0201-restricted manner. In addition, M1.1 (MUC1950-958), a promising antitumor peptide that has been tested in multiple tumors, was not able to induce stronger antitumor responses. Collectively, our results demonstrated that altered peptides from MUC1, as potential HLA-A0201-restricted CTL epitopes, could serve as peptide vaccines or constitute components of peptide-loaded dendritic cell vaccines for gastric cancer treatment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate, BioReagent, suitable for fluorescence, ≥90.0% (HPLC)
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)