Merck
  • Polyphenolic Profiling, Browning, and Glutathione Content of Sparkling Wines Produced with Nontraditional Grape Varieties: Indicator of Quality During the Biological Aging.
Polyphenolic Profiling, Browning, and Glutathione Content of Sparkling Wines Produced with Nontraditional Grape Varieties: Indicator of Quality During the Biological Aging.
Journal of food science (2019-11-12)
Saionara Sartor, Vívian Maria Burin, Nayla E Ferreira-Lima, Vinícius Caliari, Marilde T Bordignon-Luiz
ABSTRAKT
Sparkling wines were elaborated with the nontraditional varieties Villenave, Niagara, Manzoni, and Goethe, and monitored in relation to the changes in phenolic composition, browning index, and glutathione content during 18 months of biological aging (sur lies). Important changes in the phenolic profile, browning index, and glutathione content were observed in the sparkling wines during the over-lees aging period. The major phenolic compound in the sparkling wines was tyrosol, followed by caffeic, trans-caftaric, and gallic acids, catechin and epicatechin. The biological aging led to an increase in the individual phenolic compounds, especially caffeic, gallic, and ellagic acids, and an increase in the browning index was also observed during the aging period. Caffeic acid was significantly correlated with browning and aging period in all sparkling wines, which indicates that this compound can be useful as a quality marker to monitoring the biological aging profile of white sparkling wines. The results obtained indicate that the aging period (sur lie) had an important influence on the changes in the unique phenolic profile of the sparkling wines elaborated with nontraditional varieties. PRACTICAL APPLICATION: In sparkling wines production, the secondary fermentation occurring in the sealed bottle during the vinification contributes greatly to their quality and sensory complexity. The Vitis labrusca and hybrid grapes varieties represent most of the grapes cultivated in Brazil being employed in the elaboration of juices and wines. These varieties present a great oenological potential and have not been explored yet regarding to the production of white sparkling wines. The use of these nontraditional grape varieties cultivated in South Brazil may be a viable alternative in the production of white sparkling wines with biological aging potential and particular bioactive properties.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2,3-Naphthalenedicarboxaldehyde, suitable for fluorescence
Millipore
DMACA Reagent, suitable for microbiology