Merck
Identification of quality control markers in Suhuang antitussive capsule based on HPLC-PDA fingerprint and anti-inflammatory screening.
Journal of pharmaceutical and biomedical analysis (2020-01-07)
Xingdong Wu, Qinyan Liu, Dong Chen, Weiwei Qin, Bingyun Lu, Qirui Bi, Zhen Wang, Yuning Jia, Ninghua Tan
ABSTRAKT
Suhuang antitussive capsule (SH), one of traditional Chinese patent medicines, has been widely used for treating cough variant asthma and postinfectious cough in clinic. The objective of this work is to identify the characteristic and active ingredients as the quality control markers for SH based on high performance liquid chromatography with photodiode array detector (HPLC-PDA) fingerprint and screening of anti-inflammatory components. Similarity analysis (SA), hierarchical clustering analysis (HCA) and principal component analysis (PCA) were used to evaluate 16 different batches of SH. 13 compounds accounting for 36% of the total components in the fingerprint were identified and semi-quantitatively analyzed, which anti-inflammatory activity was tested with the in vitro assay. The results showed that the established chemical fingerprint could clearly distinguish different batches of SH by SA, HCA, and PCA analysis. Furthermore, four known compounds (chlorogenic acid, schisandrin, angeloylgomisin H and praeruptorin A) were screened out to be the most discriminant variables, which could be applied to quality control of SH by quantitative analysis. The semi-quantitative results showed that six compounds were major components, i.e. arctiin (10.28 ± 3.18 mg/g), ephedrine (9.26 ± 1.58 mg/g), schisandrin (3.09 ± 0.83 mg/g), pseudoephedrine (2.34 ± 1.04 mg/g), schisandrin B (1.48 ± 0.16 mg/g), and 1-caffeoylquinic acid (1.36 ± 0.42 mg/g). The anti-inflammatory results showed that SH extract, praeruptorin A, schisandrin, arctigenin and pseudoephedrine could significantly inhibit inflammatory mediator NO production in LPS-stimulated RAW264.7 macrophages. These findings indicated that praeruptorin A, schisandrin, arctiin and pseudoephedrine could be proposed as the quality control markers for SH.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction