Merck
  • Development and validation of a liquid chromatography-tandem mass spectrometric method for the quantification of 5-thio-d-glucose in rat and human plasma.
Development and validation of a liquid chromatography-tandem mass spectrometric method for the quantification of 5-thio-d-glucose in rat and human plasma.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2012-01-10)
Akiko Mizuno-Yasuhira, Shigeji Jingu, Shigeru Okuyama
ABSTRAKT
A highly selective, sensitive, and robust liquid chromatography-tandem mass spectrometric method for the determination of 5-thio-d-glucose concentrations in rat and human plasma was developed and validated. The sample preparation procedure involved protein precipitation and solid phase extraction, which efficiently removed sources of interference present in the plasma. Chromatographic separation was obtained using an NH(2)-column with distilled water and acetonitrile as the mobile phase under gradient conditions. Detection was performed using tandem mass spectrometry equipped with an electrospray ionization interface in negative ion mode. The selected reaction monitoring (SRM) transitions for 5-thio-d-glucose and an internal standard (5-thio-d-glucose-(13)C(6)) were m/z 195→m/z 105 and m/z 201→m/z 108, respectively. The correlation coefficients of the calibration curves ranged from 0.9997 to 0.9999 over a concentration range from 10 to 3000ng/mL plasma. The validated method was successfully applied to a pharmacokinetic study in rats.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
5-Thio-D-glucose, ≥98.0% (HPLC)