Merck
  • Phthalates and non-phthalate plasticizers in sediment from Korean coastal waters: Occurrence, spatial distribution, and ecological risks.
Phthalates and non-phthalate plasticizers in sediment from Korean coastal waters: Occurrence, spatial distribution, and ecological risks.
Marine pollution bulletin (2020-04-23)
Young-Sun Lee, Jae-Eun Lim, Sunggyu Lee, Hyo-Bang Moon
ABSTRAKT
Due to regulations on phthalates, non-phthalate plasticizers (NPPs) are now used as an alternative. Limited studies have been conducted on the occurrence and distribution of NPPs. In this study, sediment samples were collected from 50 locations along the Korean coast to assess the occurrence, distribution, sources, and ecological risks of phthalates and NPPs. Phthalates and NPPs were detected in all sediments, indicating ubiquitous contamination of the coastal environment. Di(2-ethylhexyl)phthalate (DEHP) and di(2-ethylhexyl)terephthalate (DEHT) were dominant, suggesting that DEHT could be an emerging contaminant of concern. The highest concentrations of phthalates and NPPs were found in sediment samples from harbors, implying they are contaminated hotspots. Sedimentary organic carbon was a major factor governing the distribution of phthalates and NPPs. Significant correlations were observed among phthalates and NPPs, suggesting similar sources and geochemical behavior. DEHP concentration exceeded threshold values, indicating potential health risks to benthic organisms in sediments.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Dihexyl phthalate, analytical standard
Sigma-Aldrich
Dess-Martin periodinane, 97%