Merck
Recent BCR stimulation induces a negative autoregulatory loop via FBXO10 mediated degradation of HGAL.
Leukemia (2019-10-02)
Fengjie Guo, Yuan Luo, Xiaoyu Jiang, XiaoQing Lu, Domenico Roberti, Chen Lossos, Kranthi Kunkalla, Marco Magistri, Lixin Rui, Ramiro Verdun, Francisco Vega, Vincent T Moy, Izidore S Lossos
ABSTRAKT
Regulating B-cell receptor (BCR) signaling after antigenic stimulation is essential to properly control immune responses. Currently known mechanisms of inhibiting BCR signaling are via co-receptor stimulation and downstream immunoreceptor tyrosine-based inhibition motif (ITIM) phosphorylation. Herein we demonstrate that BCR stimulation induces rapid and reversible palmitoylation of the SCF-FBXO10 ubiquitin E3 ligase. This results in FBXO10 relocation to the cell membrane, where it targets the human germinal center-associated lymphoma (HGAL) protein for ubiquitylation and degradation, leading to decreases in both BCR-induced calcium influx and phosphorylation of proximal BCR effectors. Importantly, FBXO10 recognition and degradation of HGAL is phosphorylation independent and instead relies on a single evolutionarily conserved HGAL amino acid residue (H91) and FBXO10 relocalization to the cytoplasmic membrane. Together our findings demonstrate the first evidence of negative BCR signaling regulation from direct BCR stimulation and define the temporospatial functions of the FBXO10-HGAL axis. FBXO10 is infrequently mutated in DLBCL but some of these mutations deregulate BCR signaling. These observations may have important implications on lymphomagenesis and other immune processes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture