Merck
Modified Potentiometric Screen-Printed Electrodes Based on Imprinting Character for Sodium Deoxycholate Determination.
Biomolecules (2020-02-12)
Ayman H Kamel, Samar Ezzat, Mona A Ahmed, Abd El-Galil E Amr, Abdulrahman A Almehizia, Mohamed A Al-Omar
ABSTRAKT
Potentiometric sensors have a great influence on the determination of most various compounds in their matrices. Therefore, efficient and new sensors were introduced to measure sodium Deoxycholate (NaDC) as a bile acid salt. These sensors are based on NaDC imprinted polymer (MIP) as sensory element. The MIP beads were synthesized using thermal polymerization pathway, in which acrylamide (AAm), ethylene glycol dimethacrylate (EGDMA), NaDC, and benzoyl peroxide (BPO) were used as the functional monomer, cross-linker, template, and initiator, respectively. The proposed sensors were fabricated using a coated screen-printed platform and the sensing membrane was modified by single-walled carbon nanotubes (SWCNTs) as an ion-to-electron transducer. The sensors exhibited high sensitivity that reached 4.7 × 10-5 M of near-Nernestian slope (-60.1 ± 0.9 mV/decade, r2 = 0.999 (n= 5)). In addition, the sensors revealed high selectivity, long lifetime, high potential stability, and conductivity that ensure reproducible and accurate results over a long time. MIP characterization was performed using Fourier Transform-Infrared (FT-IR) and a scanning electron microscope (SEM). Regarding the interaction of NaDC with serum albumin (SA), albumin is determined in human serum samples as human serum albumin (HSA), which was collected from different volunteers of different ages and gender.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor