Merck
  • A 1-week extension of a ketogenic diet provides a further decrease in myocardial 18F-FDG uptake and a high detectability of myocarditis with FDG-PET.
A 1-week extension of a ketogenic diet provides a further decrease in myocardial 18F-FDG uptake and a high detectability of myocarditis with FDG-PET.
Journal of nuclear cardiology : official publication of the American Society of Nuclear Cardiology (2018-08-22)
Alexandra Clément, Henri Boutley, Sylvain Poussier, Julien Pierson, Mickael Lhuillier, Allan Kolodziej, Jean-Luc Olivier, Gilles Karcher, Pierre-Yves Marie, Fatiha Maskali
ABSTRAKT
Short periods of fasting and/or low-carbohydrate diet have been proven beneficial for decreasing the myocardial uptake of 18F-fluorodeoxyglucose (18F-FDG) and enhancing the detection of inflammatory heart diseases by 18F-FDG positron emission tomography (PET). This study aimed at determining whether this benefit is increased when a low-carbohydrate ketogenic diet is prolonged up to 7 days. Wistar rats underwent serial 18F-FDG-PET imaging after an 18-hour fasting period and after 2, 4 and 7 days of a ketogenic diet (3% carbohydrate) and they were compared to rats submitted to the same protocol but with normal diet (44% carbohydrate). The 18F-FDG-PET/ketogenic protocol was also applied in rats with immune myocarditis (injection of porcine cardiac myosin). The 7-day ketogenic diet was associated with (1) a sustained increase in circulating ketone bodies at an equivalent level to that reached after 18-hour fasting, (2) a gradual decrease in 18F-FDG uptake within normal myocardium reaching a lower level compared to fasting at the 7th day (myocardium-to-blood ratios: 1.68 ± 1.02 vs 3.25 ± 1.40, P < .05) and (3) a high 18F-FDG-PET detectability of myocarditis areas. One-week extension of a ketogenic diet provides a further decrease in the 18F-FDG uptake of normal myocardium and a high detectability of inflammatory areas.