Merck
The solid-state structure of the β-blocker metoprolol: a combined experimental and in silico investigation.
Acta crystallographica. Section C, Structural chemistry (2019-02-06)
Patrizia Rossi, Paola Paoli, Laura Chelazzi, Luca Conti, Andrea Bencini
ABSTRAKT
Metoprolol {systematic name: (RS)-1-isopropylamino-3-[4-(2-methoxyethyl)phenoxy]propan-2-ol}, C15H25NO3, is a cardioselective β1-adrenergic blocking agent that shares part of its molecular skeleton with a large number of other β-blockers. Results from its solid-state characterization by single-crystal and variable-temperature powder X-ray diffraction and differential scanning calorimetry are presented. Its molecular and crystal arrangements have been further investigated by molecular modelling, by a Cambridge Structural Database (CSD) survey and by Hirshfeld surface analysis. In the crystal, the side arm bearing the isopropyl group, which is common to other β-blockers, adopts an all-trans conformation, which is the most stable arrangement from modelling data. The crystal packing of metoprolol is dominated by an O-H...N/N...H-O pair of hydrogen bonds (as also confirmed by a Hirshfeld surface analysis), which gives rise to chains containing alternating R and S metoprolol molecules extending along the b axis, supplemented by a weaker O...H-N/N-H...O pair of interactions. In addition, within the same stack of molecules, a C-H...O contact, partially oriented along the b and c axes, links homochiral molecules. Amongst the solid-state structures of molecules structurally related to metoprolol deposited in the CSD, the β-blocker drug betaxolol shows the closest analogy in terms of three-dimensional arrangement and interactions. Notwithstanding their close similarity, the crystal lattices of the two drugs respond differently on increasing temperature: metoprolol expands anisotropically, while for betaxolol, an isotropic thermal expansion is observed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Betaxolol hydrochloride, >98% (HPLC)