Merck
  • Human T lymphocyte mitogenesis in response to the B oligomer of pertussis toxin is associated with an early elevation in cytosolic calcium concentrations.
Human T lymphocyte mitogenesis in response to the B oligomer of pertussis toxin is associated with an early elevation in cytosolic calcium concentrations.
FEBS letters (1987-12-10)
C F Strnad, R A Carchman
ABSTRAKT
Pertussis toxin was found to serve as a mitogen in the human T lymphocyte, an effect which could be mimicked by its resolved binding component, the B oligomer. The mechanism of action of this component appeared to involve a rapid and sustained elevation of cytosolic calcium levels, as monitored by fura-2 fluorescence. The source of mobilized calcium was predominantly extracellular, suggesting that the binding of the B oligomer to the T cell plasma membrane in some way elicited calcium channel activation. Notably, the influx of calcium was not observed with cholera toxin, an AB toxin lacking mitogenic effects on the human T lymphocyte.