Merck
  • Synthetic thyrotropin-releasing hormone. Activity in men and women, specificity of action, inhibition by triiodothyronine, and activity orally.
Synthetic thyrotropin-releasing hormone. Activity in men and women, specificity of action, inhibition by triiodothyronine, and activity orally.
Journal of medicinal chemistry (1971-06-01)
C Y Bowers, A V Schally, A Kastin, A Arimura, D S Schalch, C Gual, E Castineda, K Folkers