Merck
Mitochondrial peptide BRAWNIN is essential for vertebrate respiratory complex III assembly.
Nature communications (2020-03-13)
Shan Zhang, Boris Reljić, Chao Liang, Baptiste Kerouanton, Joel Celio Francisco, Jih Hou Peh, Camille Mary, Narendra Suhas Jagannathan, Volodimir Olexiouk, Claire Tang, Gio Fidelito, Srikanth Nama, Ruey-Kuang Cheng, Caroline Lei Wee, Loo Chien Wang, Paula Duek Roggli, Prabha Sampath, Lydie Lane, Enrico Petretto, Radoslaw M Sobota, Suresh Jesuthasan, Lisa Tucker-Kellogg, Bruno Reversade, Gerben Menschaert, Lei Sun, David A Stroud, Lena Ho
ABSTRAKT
The emergence of small open reading frame (sORF)-encoded peptides (SEPs) is rapidly expanding the known proteome at the lower end of the size distribution. Here, we show that the mitochondrial proteome, particularly the respiratory chain, is enriched for small proteins. Using a prediction and validation pipeline for SEPs, we report the discovery of 16 endogenous nuclear encoded, mitochondrial-localized SEPs (mito-SEPs). Through functional prediction, proteomics, metabolomics and metabolic flux modeling, we demonstrate that BRAWNIN, a 71 a.a. peptide encoded by C12orf73, is essential for respiratory chain complex III (CIII) assembly. In human cells, BRAWNIN is induced by the energy-sensing AMPK pathway, and its depletion impairs mitochondrial ATP production. In zebrafish, Brawnin deletion causes complete CIII loss, resulting in severe growth retardation, lactic acidosis and early death. Our findings demonstrate that BRAWNIN is essential for vertebrate oxidative phosphorylation. We propose that mito-SEPs are an untapped resource for essential regulators of oxidative metabolism.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Tubulin, Polyglutamylated antibody, Mouse monoclonal, clone B3, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-UQCRB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution