Merck
Gas7 knockout affects PINK1 expression and mitochondrial dynamics in mouse cortical neurons.
FASEB bioAdvances (2020-03-13)
Jagannatham Naidu Bhupana, Bo-Tsang Huang, Gunn-Guang Liou, Marcus J Calkins, Sue Lin-Chao
ABSTRAKT
Dynamic fission and fusion events regulate mitochondrial shape, distribution, and rejuvenation, and proper control of these processes is essential for neuronal homeostasis. Here, we report that Gas7, a known cytoskeleton regulator, controls mitochondrial dynamics within neurons of the central nervous system. In this study, we generated an improved Gas7-knockout mouse and evaluated its mitochondrial phenotype. We first identified Gas7 in mitochondrial fractions from wild-type brain tissue, and observed Gas7 colocalization with mitochondria in primary cortical neurons. In Gas7-deficient brain tissue and neuronal cultures mitochondria were elongated with perinuclear clustering. These morphological abnormalities were associated with increased levels mitochondrial fusion proteins and increased PKA-dependent phosphorylation of Drp-1 in brain tissues, suggesting an imbalance of mitochondrial fusion and fission. Moreover, expression of mitochondrial quality control kinase, PINK1, and PINK1-specific phosphorylation of Mfn-2 (S442), Parkin (S65), and ubiquitin (S65) were all reduced in the knockout cells. Ectopic expression of Gas7 restored mitochondrial morphology and distribution, as well as PINK1 expression in Gas7-null cortical neurons. Collectively, our results introduce a novel role of mouse Gas7 in determining the dynamics, morphology, and intracellular distribution of neuronal mitochondria, which are expected to be required for normal neuronal function.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-FIS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-phospho-PDHE1-A type I (Ser293) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-GAPDH Antibody, from chicken, purified by affinity chromatography