- Structure of a molluscan cardioexcitatory neuropeptide.
Structure of a molluscan cardioexcitatory neuropeptide.
Science (New York, N.Y.) (1977-08-12)
D A Price, M J Greenberg
PMID877582
ABSTRAKT
A cardioexcitatory substance from ganglia of the clam Macrocallista nimbosa, formerly designated peak C, is the tetrapeptide amide Phe-Met-Arg-Phe-NH2. Its structure was determined by the combined use of Edman dansyl degradation and tryptic digestion. The structure was confirmed by synthesis. This neuropeptide is active at about 10(-8)M when assayed on molluscan muscle.