Merck
Neuronal BIN1 Regulates Presynaptic Neurotransmitter Release and Memory Consolidation.
Cell reports (2020-03-12)
Pierre De Rossi, Toshihiro Nomura, Robert J Andrew, Nicolas Y Masse, Vandana Sampathkumar, Timothy F Musial, Ari Sudwarts, Aleksandra J Recupero, Thomas Le Metayer, Mitchell T Hansen, Ha-Na Shim, Sofia V Krause, David J Freedman, Vytas P Bindokas, Narayanan Kasthuri, Daniel A Nicholson, Anis Contractor, Gopal Thinakaran
ABSTRAKT
BIN1, a member of the BAR adaptor protein family, is a significant late-onset Alzheimer disease risk factor. Here, we investigate BIN1 function in the brain using conditional knockout (cKO) models. Loss of neuronal Bin1 expression results in the select impairment of spatial learning and memory. Examination of hippocampal CA1 excitatory synapses reveals a deficit in presynaptic release probability and slower depletion of neurotransmitters during repetitive stimulation, suggesting altered vesicle dynamics in Bin1 cKO mice. Super-resolution and immunoelectron microscopy localizes BIN1 to presynaptic sites in excitatory synapses. Bin1 cKO significantly reduces synapse density and alters presynaptic active zone protein cluster formation. Finally, 3D electron microscopy reconstruction analysis uncovers a significant increase in docked and reserve pools of synaptic vesicles at hippocampal synapses in Bin1 cKO mice. Our results demonstrate a non-redundant role for BIN1 in presynaptic regulation, thus providing significant insights into the fundamental function of BIN1 in synaptic physiology relevant to Alzheimer disease.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Glutamate Receptor 1 Antibody, phosphoSer 831, Chemicon®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glutamate Receptor 1 Antibody, phosphoSer 845, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-MAP2 antibody produced in mouse, ascites fluid, clone HM-2
Sigma-Aldrich
Monoclonal Anti-Synaptophysin antibody produced in mouse, clone SVP-38, ascites fluid
Sigma-Aldrich
Anti-CNPase Antibody, clone 11-5B, clone 11-5B, Chemicon®, from mouse