Merck
  • Orphan Nuclear Receptor RORα Regulates Enzymatic Metabolism of Cerebral 24S-Hydroxycholesterol through CYP39A1 Intronic Response Element Activation.
Orphan Nuclear Receptor RORα Regulates Enzymatic Metabolism of Cerebral 24S-Hydroxycholesterol through CYP39A1 Intronic Response Element Activation.
International journal of molecular sciences (2020-05-13)
Hiroshi Matsuoka, Miyu Katayama, Ami Ohishi, Kaoruko Miya, Riki Tokunaga, Sou Kobayashi, Yuya Nishimoto, Kazutake Hirooka, Akiho Shima, Akihiro Michihara
ABSTRAKT
Oxysterols, important regulators of cholesterol homeostasis in the brain, are affected by neurodegenerative diseases. Early-onset Alzheimer's disease is associated with higher levels of circulating brain-derived 24S-hydroxycholesterol (24S-OHC). Conversion of cholesterol to 24S-OHC is mediated by cholesterol 24S-hydroxylase in the brain, which is the major pathway for oxysterol elimination, followed by oxidation through hepatic first-pass metabolism by CYP39A1. Abnormal CYP39A1 expression results in accumulation of 24S-OHC, influencing neurodegenerative disease-related deterioration; thus, it is important to understand the normal elimination of 24S-OHC and the system regulating CYP39A1, a selective hepatic metabolic enzyme of 24S-OHC. We examined the role of transcriptional regulation by retinoic acid receptor-related orphan receptor α (RORα), a nuclear receptor that responds to oxysterol ligands. In humans, the promoter and first intronic regions of CYP39A1 contain two putative RORα response elements (ROREs). RORα binding and responses of these ROREs were assessed using electrophoretic mobility shift, chromatin immunoprecipitation, and luciferase reporter assays. CYP39A1 was upregulated by RORα overexpression in HEK293 cells, while RORα knockdown by siRNA significantly downregulated CYP39A1 expression in human hepatoma cells. Additionally, CYP39A1 was induced by RORα agonist treatment, suggesting that CYP39A1 expression is activated by RORα nuclear receptors. This may provide a way to increase CYP39A1 activity using RORα agonists, and help halt 24S-OHC accumulation in neurodegenerative illnesses.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Cytochrome P450 39A1 antibody produced in rabbit, affinity isolated antibody