Merck
Small extracellular vesicles deliver miR-21 and miR-217 as pro-senescence effectors to endothelial cells.
Journal of extracellular vesicles (2020-03-12)
Emanuela Mensà, Michele Guescini, Angelica Giuliani, Maria Giulia Bacalini, Deborah Ramini, Giacomo Corleone, Manuela Ferracin, Gianluca Fulgenzi, Laura Graciotti, Francesco Prattichizzo, Leonardo Sorci, Michela Battistelli, Vladia Monsurrò, Anna Rita Bonfigli, Maurizio Cardelli, Rina Recchioni, Fiorella Marcheselli, Silvia Latini, Serena Maggio, Mirco Fanelli, Stefano Amatori, Gianluca Storci, Antonio Ceriello, Vilberto Stocchi, Maria De Luca, Luca Magnani, Maria Rita Rippo, Antonio Domenico Procopio, Claudia Sala, Iva Budimir, Cristian Bassi, Massimo Negrini, Paolo Garagnani, Claudio Franceschi, Jacopo Sabbatinelli, Massimiliano Bonafè, Fabiola Olivieri
ABSTRAKT
The role of epigenetics in endothelial cell senescence is a cutting-edge topic in ageing research. However, little is known of the relative contribution to pro-senescence signal propagation provided by microRNAs shuttled by extracellular vesicles (EVs) released from senescent cells. Analysis of microRNA and DNA methylation profiles in non-senescent (control) and senescent (SEN) human umbilical vein endothelial cells (HUVECs), and microRNA profiling of their cognate small EVs (sEVs) and large EVs demonstrated that SEN cells released a significantly greater sEV number than control cells. sEVs were enriched in miR-21-5p and miR-217, which target DNMT1 and SIRT1. Treatment of control cells with SEN sEVs induced a miR-21/miR-217-related impairment of DNMT1-SIRT1 expression, the reduction of proliferation markers, the acquisition of a senescent phenotype and a partial demethylation of the locus encoding for miR-21. MicroRNA profiling of sEVs from plasma of healthy subjects aged 40-100 years showed an inverse U-shaped age-related trend for miR-21-5p, consistent with senescence-associated biomarker profiles. Our findings suggest that miR-21-5p/miR-217 carried by SEN sEVs spread pro-senescence signals, affecting DNA methylation and cell replication.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Calnexin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution