Merck
Phytanoyl-coenzyme A hydroxylase deficiency -- the enzyme defect in Refsum's disease.
The New England journal of medicine (1997-07-10)
G A Jansen, R J Wanders, P A Watkins, S J Mihalik
MATERIAŁY
Numer produktu
Marka
Opis produktu
Avanti
4ME 16:0 Coenzyme A, Avanti Research - A Croda Brand 870742P, powder