Merck
A rapid fluorescence assay for sphingosine-1-phosphate lyase enzyme activity.
Journal of lipid research (2007-09-18)
Padmavathi Bandhuvula, Henrik Fyrst, Julie D Saba
ABSTRAKT
Sphingosine-1-phosphate (S1P) lyase (SPL) catalyzes the conversion of S1P to ethanolamine phosphate and hexadecenal. This enzyme plays diverse roles in physiology and disease and, thus, may be useful as a disease marker and/or drug target. Unfortunately, the radioisotope-based assay currently used to quantify SPL activity is suboptimal. We have devised an assay using a commercially available omega(7-nitro-2-1,3-benzoxadiazol-4-yl)-d-erythro (NBD)-labeled fluorescent substrate. Alternatively, we provide a method for synthesis of the substrate from NBD-sphingosine. Enzyme activity is determined by following the formation of NBD-aldehyde product, which is isolated from unreacted substrate by lipid extraction and quantified after separation by HPLC using a C18 column. A fluorescent NBD-C18-sphingosine internal standard is used to control for extraction efficiency. The reaction is linear over 20 min and total protein concentrations of 20-200 mg/l. The sensitivity of the fluorescence assay is comparable to or better than that of the radioactive assay, and SPL levels as low as 8 pmol/mg/min were readily detected. Semicarbazide, a nonspecific SPL inhibitor, reduced SPL activity in vitro by approximately 70% using both standard and fluorescence methods. Product inhibition was not observed using ethanolamine phosphate and a commercially available source of hexadecenal. This method is suitable for quantifying SPL activity in a variety of cell and tissue sources.