Merck
  • Effective production of chitinase and chitosanase byStreptomyces griseus HUT 6037 using colloidal chitin and various degrees of deacetylation of chitosan
Effective production of chitinase and chitosanase byStreptomyces griseus HUT 6037 using colloidal chitin and various degrees of deacetylation of chitosan
Biotechnology and Bioprocess Engineering: BBE (1999)
Jung, H., et al.