- C/EBPα Induces Highly Efficient Macrophage Transdifferentiation of B Lymphoma and Leukemia Cell Lines and Impairs Their Tumorigenicity.
C/EBPα Induces Highly Efficient Macrophage Transdifferentiation of B Lymphoma and Leukemia Cell Lines and Impairs Their Tumorigenicity.
Cell reports (2017-05-13)
Francesca Rapino, Eloy F Robles, Jose A Richter-Larrea, Eric M Kallin, Jose A Martinez-Climent, Thomas Graf
PMID28494875
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
BLaER1 Human B-cell Precursor Leukemia Cell Line, BLaER1 human B-cell precursor leukemia cell line may be transdifferentiated and used to model human monocytes.