Merck
Epigenetic dynamics of imprinted X inactivation during early mouse development.
Science (New York, N.Y.) (2003-12-13)
Ikuhiro Okamoto, Arie P Otte, C David Allis, Danny Reinberg, Edith Heard
ABSTRAKT
The initiation of X-chromosome inactivation is thought to be tightly correlated with early differentiation events during mouse development. Here, we show that although initially active, the paternal X chromosome undergoes imprinted inactivation from the cleavage stages, well before cellular differentiation. A reversal of the inactive state, with a loss of epigenetic marks such as histone modifications and polycomb proteins, subsequently occurs in cells of the inner cell mass (ICM), which give rise to the embryo-proper in which random X inactivation is known to occur. This reveals the remarkable plasticity of the X-inactivation process during preimplantation development and underlines the importance of the ICM in global reprogramming of epigenetic marks in the early embryo.