Merck
  • Functional Annotations of Single-Nucleotide Polymorphism (SNP)-Based and Gene-Based Genome-Wide Association Studies Show Genes Affecting Keratitis Susceptibility.
Functional Annotations of Single-Nucleotide Polymorphism (SNP)-Based and Gene-Based Genome-Wide Association Studies Show Genes Affecting Keratitis Susceptibility.
Medical science monitor : international medical journal of experimental and clinical research (2020-05-26)
Yue Xu, Xiao-Lin Yang, Xiao-Long Yang, Ya-Ru Ren, Xin-Yu Zhuang, Lei Zhang, Xiao-Feng Zhang
ABSTRAKT
BACKGROUND Keratitis is a complex condition in humans and is the second most common cause of legal blindness worldwide. MATERIAL AND METHODS To reveal the genomic loci underlying keratitis, we performed functional annotations of SNP-based and gene-based genome-wide association studies of keratitis in the UK Biobank (UKB) cohort with 337 199 subjects of European ancestry. RESULTS The publicly available SNP-based association results showed a total of 34 SNPs, from 14 distinct loci, associated with keratitis in the UKB. Gene-based association analysis identified 2 significant genes: IQCF3 (p=2.0×10⁻⁶) and SOD3 (p=2.0×10⁻⁶). Thirty-two candidate genes were then prioritized using information from multiple sources. The overlap of IQCF3 in these 2 analyses resulted in a total of 33 hub genes. Functional annotation of hub genes was performed and transcriptional factors of IQCF3 and SOD3 were predicted. CONCLUSIONS A total of 34 SNPs from 14 distinct loci were identified as being associated with keratitis, and 32 candidate genes were then prioritized. In addition, IQCF3 and SOD3 were identified by their p values through gene-based tests on the basis of individual SNP-based tests. The functional relationship between these suspect genes and keratitis suggest that IQCF3 and SOD3 are candidate genes underlying keratitis.