Merck
Enhanced gastrointestinal passive paracellular permeability contributes to the obesity-associated hyperoxaluria.
American journal of physiology. Gastrointestinal and liver physiology (2018-10-12)
Mohamed Bashir, Jon Meddings, Altayeb Alshaikh, Daniel Jung, Kim Le, Ruhul Amin, Sireesha Ratakonda, Sapna Sharma, Ignacio Granja, Mustafa Satti, John Asplin, Hatim Hassan
ABSTRAKT
Most kidney stones (KS) are composed of calcium oxalate and small increases in urine oxalate enhance the stone risk. Obesity is a risk factor for KS, and urinary oxalate excretion increases with increased body size. We previously established the obese ob/ob ( ob) mice as a model (3.3-fold higher urine oxalate) to define the pathogenesis of obesity-associated hyperoxaluria (OAH). The purpose of this study was to test the hypothesis that the obesity-associated enhanced small intestinal paracellular permeability contributes to OAH by increasing passive paracellular intestinal oxalate absorption. ob Mice have significantly higher jejunal (1.6-fold) and ileal (1.4-fold) paracellular oxalate absorption ex vivo and significantly higher (5-fold) urine [13C]oxalate following oral gavage with [13C]oxalate, indicating increased intestinal oxalate absorption in vivo. The observation of higher oxalate absorption in vivo compared with ex vivo suggests the possibility of increased paracellular permeability along the entire gut. Indeed, ob mice have significantly higher fractions of the administered sucrose (1.7-fold), lactulose (4.4-fold), and sucralose (3.1-fold) excreted in the urine, reflecting increased gastric, small intestinal, and colonic paracellular permeability, respectively. The ob mice have significantly reduced gastrointestinal occludin, zonula occludens-1, and claudins-1 and -3 mRNA and total protein expression. Proinflammatory cytokines and oxidative stress, which are elevated in obesity, significantly enhanced paracellular intestinal oxalate absorption in vitro and ex vivo. We conclude that obese mice have significantly higher intestinal oxalate absorption and enhanced gastrointestinal paracellular permeability in vivo, which would likely contribute to the pathogenesis of OAH, since there is a transepithelial oxalate concentration gradient to drive paracellular intestinal oxalate absorption. NEW & NOTEWORTHY This study shows that the obese ob/ob mice have significantly increased gastrointestinal paracellular oxalate absorption and remarkably enhanced paracellular permeability along the entire gut in vivo, which are likely mediated by the obesity-associated increased systemic and intestinal inflammation and oxidative stress. A transepithelial oxalate concentration gradient driving gastrointestinal paracellular oxalate absorption exists, and therefore, our novel findings likely contribute to the hyperoxaluria observed in the ob/ob mice and hence to the pathogenesis of obesity-associated hyperoxaluria.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture