Merck
Sugar Alcohols of Polyol Pathway Serve as Alarmins to Mediate Local-Systemic Innate Immune Communication in Drosophila.
Cell host & microbe (2019-07-28)
Shuo Yang, Yaya Zhao, Junjing Yu, Zhiqin Fan, Si-Tang Gong, Hong Tang, Lei Pan
ABSTRAKT
Interorgan immunological communication is critical to connect the local-systemic innate immune response and orchestrate a homeostatic host defense. However, the factors and their roles in this process remain unclear. We find Drosophila IMD response in guts can sequentially trigger a systemic IMD reaction in the fat body. Sugar alcohols of the polyol pathway are essential for the spatiotemporal regulation of gut-fat body immunological communication (GFIC). IMD activation in guts causes elevated levels of sorbitol and galactitol in hemolymph. Aldose reductase (AR) in hemocytes, the rate-limiting enzyme of the polyol pathway, is necessary and sufficient for the increase of plasma sugar alcohols. Sorbitol relays GFIC by subsequent activation of Metalloprotease 2, which cleaves PGRP-LC to activate IMD response in fat bodies. Thus, this work unveils how GFIC relies on the intermediate activation of the polyol pathway in hemolymph and demonstrates that AR provides a critical metabolic checkpoint in the global inflammatory response.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Glucose (HK) Assay Kit, sufficient for 20 assays
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Roche
cOmplete, Mini, Koktajl inhibitorów proteaz bez EDTA, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial