Merck
Steroidogenic acute regulatory protein (StAR) is a sterol transfer protein.
The Journal of biological chemistry (1998-10-03)
C B Kallen, J T Billheimer, S A Summers, S E Stayrook, M Lewis, J F Strauss
ABSTRAKT
Steroidogenic acute regulatory protein (StAR) plays a critical role in steroidogenesis by enhancing the delivery of substrate cholesterol from the outer mitochondrial membrane to the cholesterol side chain cleavage enzyme system on the inner membrane. A recombinant StAR protein lacking the first N-terminal 62 amino acid residues that includes the mitochondrial targeting sequence was shown to stimulate the transfer of cholesterol and beta-sitosterol from liposomes to heat-treated mitochondria in a dose-, time-, and temperature-dependent manner. A recombinant mutant StAR protein that cannot stimulate steroidogenesis by isolated mitochondria did not promote sterol transfer. Unlike the more promiscuous lipid transfer protein, sterol carrier protein 2 (SCP2), StAR did not stimulate phosphatidylcholine transfer in our assay system. The recombinant StAR protein increased cholesterol transfer to heat-treated microsomes as well as to heat- and trypsin-treated mitochondria. These observations demonstrate that StAR has sterol transfer activity, which may reflect an ability to enhance desorption of cholesterol from sterol-rich donor membranes. We suggest that the ability of StAR to promote sterol transfer explains its steroidogenic activity.