Merck
Proanthocyanidin promotes functional recovery of spinal cord injury via inhibiting ferroptosis.
Journal of chemical neuroanatomy (2020-06-01)
Huangao Zhou, Chaoyun Yin, Zhiming Zhang, Haowen Tang, Wen Shen, Xuan Zha, Ming Gao, Jifu Sun, Xiuquan Xu, Qian Chen
ABSTRAKT
Improving the microenvironment of lesioned spinal cord to minimize the secondary injury is one important strategy to treat spinal cord injury (SCI). The ensuing hemorrhage after SCI has tight connection with ferroptosis. This study investigated the effects of proanthocyanidins (PACs) on SCI repair and the underlying mechanisms. Adult female mice were divided into four groups, including sham, SCI, PACs5 and PACs10 (i.p. 5 and 10 mg/kg PACs after SCI respectively). The impacts of SCI and PACs treatment on redox parameters (iron contents, TBARS, GSH, and GPX activities) and ferroptosis essential factors such as ACSL4, LPCAT3, Alox15B, Nrf2, HO-1, GPX4 were investigated. The results demonstrated that PACs treatment significantly decreased the levels of iron, TBARS, ACSL4, and Alox15B, while increased the levels of GSH, GPX4, Nrf2, and HO-1 in traumatic spinal cords. Above all, PACs improved the locomotive function of SCI mice. These results suggest that PACs might be potential therapeutics for SCI repair by inhibiting ferroptosis in SCI.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Triton X-100, laboratory grade