Merck
Klotho is regulated by transcription factor Sp1 in renal tubular epithelial cells.
BMC molecular and cell biology (2020-06-24)
Yan Li, Yong Liu, Kailong Wang, Yinghui Huang, Wenhao Han, Jiachuan Xiong, Ke Yang, Mingying Liu, Tangli Xiao, Chi Liu, Ting He, Xianjin Bi, Jingbo Zhang, Bo Zhang, Jinghong Zhao
ABSTRAKT
Klotho is a multifunctional protein, which exists both in a membrane bound and a soluble form. In renal tubules, Klotho is involved in cell senescence, anti-oxidant response, and renal fibrosis, thus regulation of its expression is critical to understand its roles in renal diseases. Indeed, reduced expression was observed in various renal disease. However, the mechanisms underlying transcriptional regulation of the human klotho gene (KL) largely remain unknown. Here we demonstrated that the Klotho expression in human renal tubular epithelial cells (RTECs) was enhanced by overexpression of the transcription factor Sp1. On the contrary, Klotho expression was decreased by Sp1 knockdown. Besides, increased expression of Sp1 alleviated TGF-β1-induced fibrosis in HK-2 cells by inducing Klotho expression. Luciferase reporter assays and chromatin immunoprecipitation assays further identified the binding site of Sp1 was located in - 394 to - 289 nt of the KL promoter, which was further confirmed by mutation analysis. These data demonstrate that KL is a transcriptional target of Sp1 and TGF-β1-induced fibrosis was alleviated by Sp1 in human RTECs by directly modulating Klotho expression, which help to further understand the transcriptional regulation of Klotho in renal disease models.