Merck
Identifying Athlete Body Fluid Changes During a Competitive Season With Bioelectrical Impedance Vector Analysis.
International journal of sports physiology and performance (2019-06-13)
Francesco Campa, Catarina N Matias, Elisabetta Marini, Steven B Heymsfield, Stefania Toselli, Luís B Sardinha, Analiza M Silva
ABSTRAKT
To analyze the association between body fluid changes evaluated by bioelectrical impedance vector analysis and dilution techniques over a competitive season in athletes. A total of 58 athletes of both sexes (men: age 18.7 [4.0] y and women: age 19.2 [6.0] y) engaging in different sports were evaluated at the beginning (pre) and 6 months after (post) the competitive season. Deuterium dilution and bromide dilution were used as the criterion methods to assess total body water (TBW) and extracellular water (ECW), respectively; intracellular water (ICW) was calculated as TBW-ECW. Bioelectrical resistance and reactance were obtained with a phase-sensitive 50-kHz bioelectrical impedance analysis device; bioelectrical impedance vector analysis was applied. Dual-energy X-ray absorptiometry was used to assess fat mass and fat-free mass. The athletes were empirically classified considering TBW change (pre-post, increase or decrease) according to sex. Significant mean vector displacements in the postgroups were observed in both sexes. Specifically, reductions in vector length (Z/H) were associated with increases in TBW and ICW (r = -.718, P < .01; r = -.630, P < .01, respectively) and decreases in ECW:ICW ratio (r = .344, P < .05), even after adjusting for age, height, and sex. Phase-angle variations were positively associated with TBW and ICW (r = .458, P < .01; r = .564, P < .01, respectively) and negatively associated with ECW:ICW (r = -.436, P < .01). Phase angle significantly increased in all the postgroups except in women in whom TBW decreased. The results suggest that bioelectrical impedance vector analysis is a suitable method to obtain a qualitative indication of body fluid changes during a competitive season in athletes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
BIO-Acetoxime, ≥98% (HPLC)