Merck
Elucidation of anti-HIV mechanism of sulfated cellobiose-polylysine dendrimers.
Carbohydrate research (2020-07-14)
Song Weiyue, Li Ying, Taisei Kanamoto, Daisuke Asai, Hiromu Takemura, Hideki Nakashima, Kensuke Miyazaki, Takashi Yoshida
ABSTRAKT
Three new spherical sulfated cellobiose-polylysine dendrimers of increasing generations bearing negatively charged sulfate groups were prepared by sulfating the corresponding cellobiose-polylysine dendrimers. The first, second, and third-generation derivatives exhibited potent anti-HIV activity with EC50 values of 3.7, 0.6, and 1.5 μg/mL, respectively, in constant to sulfated oligosaccharides with low anti-HIV activity, while the second-generation sulfated dendrimer was the most active. Surface plasmon resonance measurements with poly-l-lysine bearing positively charged amino acids as a model of the HIV surface glycoprotein gp120, indicated that the second-generation dendrimer had the lowest dissociation constant (KD = 1.86 × 10-12 M). Both the particle size and ζ potential increased in the presence of poly-l-lysine. It was proven that the moderate distance between the terminal sulfated cellobiose units in the second-generation dendrimer favored the high anti-HIV activity, owing to the electrostatic interactions developed due to the cluster effect.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O